Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111105.3 | NP_186887.1 | 1 - 2 | 1529 | 78 | |
GenBank mRNA | AF370149.1 | - | 1 - 2 | 751 | 78 | |
AY059092.1 | - | 1 - 2 | 686 | 78 | ||
AY088779.1 | - | 1 - 2 | 756 | 78 | ||
L81120.1 | - | 1 - 2 | 710 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |