Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202864.1 | NP_001189793.1 | 10 - 11 | 1225 | 88 | |
NM_111121.4 | NP_186903.1 | 10 - 11 | 1322 | 88 | ||
GenBank mRNA | AB109196.1 | - | 10 - 11 | 1059 | 88 | |
AY039562.1 | - | 10 - 11 | 1189 | 88 | ||
AY113034.1 | - | 10 - 11 | 1059 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |