Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202864.1 | NP_001189793.1 | 3 - 4 | 454 | 73 | |
NM_111121.4 | NP_186903.1 | 3 - 4 | 548 | 73 | ||
NM_202482.3 | NP_974211.1 | 3 - 4 | 476 | 73 | ||
GenBank mRNA | AB109196.1 | - | 3 - 4 | 285 | 73 | |
AY039562.1 | - | 3 - 4 | 415 | 73 | ||
AY113034.1 | - | 3 - 4 | 285 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |