Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111125.4 | NP_186906.1 | 3 - 4 | 842 | 64 | |
GenBank mRNA | AY070447.1 | - | 3 - 4 | 834 | 64 | |
AY096564.1 | - | 3 - 4 | 718 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |