Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111175.3 | NP_186955.1 | 3 - 4 | 2261 | 86 | |
GenBank mRNA | AF232907.1 | - | 3 - 4 | 1939 | 86 | |
AF360180.1 | - | 3 - 4 | 1943 | 86 | ||
AY039996.1 | - | 2 - 3 | 1691 | 86 | ||
KJ138844.1 | - | 2 - 3 | 1691 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |