Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084632.1 | NP_001078101.1 | 13 - 14 | 2178 | 114 | |
NM_001202869.2 | NP_001189798.1 | 16 - 17 | 2541 | 114 | ||
NM_001337464.1 | NP_001327050.1 | 10 - 11 | 1659 | 114 | ||
NM_111200.5 | NP_566199.4 | 17 - 18 | 2783 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |