Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111235.4 | NP_187014.1 | 9 - 10 | 1168 | 72 | |
GenBank mRNA | AF082157.1 | - | 9 - 10 | 1132 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |