Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK-HSDH II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118091.3 | NP_193706.2 | 2 - 3 | 383 | 82 | |
NM_202847.3 | NP_974576.1 | 2 - 3 | 424 | 82 | ||
GenBank mRNA | AK228252.1 | - | 2 - 3 | 352 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |