Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036597.2 | NP_001031674.1 | 5 - 6 | 1070 | 102 | |
NM_118120.5 | NP_567593.1 | 5 - 6 | 1118 | 102 | ||
GenBank mRNA | BT026461.1 | - | 5 - 6 | 925 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |