Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341398.1 | NP_001329687.1 | 7 - 8 | 993 | 133 | |
NM_118126.4 | NP_193740.1 | 7 - 8 | 997 | 133 | ||
GenBank mRNA | BT025334.1 | - | 7 - 8 | 831 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |