Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341459.1 | NP_001328972.1 | 5 - 6 | 951 | 78 | |
NM_118205.2 | NP_193819.1 | 5 - 6 | 662 | 78 | ||
GenBank mRNA | AY044315.1 | - | 4 - 5 | 444 | 78 | |
AY058151.1 | - | 5 - 6 | 611 | 78 | ||
AY101538.1 | - | 4 - 5 | 444 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |