Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341460.1 | NP_001328331.1 | 2 - 3 | 420 | 118 | |
NM_118208.1 | NP_193822.1 | 2 - 3 | 420 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |