Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203853.2 | NP_001190782.1 | 6 - 7 | 2094 | 91 | |
NM_118209.3 | NP_567616.1 | 5 - 6 | 2239 | 91 | ||
GenBank mRNA | AF327068.1 | - | 5 - 6 | 2021 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |