Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118225.5 | NP_193839.4 | 13 - 14 | 2702 | 97 | |
GenBank mRNA | AF515728.1 | - | 13 - 14 | 2569 | 97 | |
AY081328.1 | - | 13 - 14 | 2615 | 97 | ||
BT010547.1 | - | 13 - 14 | 2519 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |