Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118225.5 | NP_193839.4 | 8 - 9 | 2007 | 106 | |
GenBank mRNA | AF515728.1 | - | 8 - 9 | 1874 | 106 | |
AY081328.1 | - | 8 - 9 | 1920 | 106 | ||
BT010547.1 | - | 8 - 9 | 1824 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |