Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118235.4 | NP_567620.1 | 7 - 8 | 841 | 77 | |
NM_179085.3 | NP_849416.1 | 7 - 8 | 798 | 77 | ||
NM_202852.2 | NP_974581.1 | 7 - 8 | 959 | 77 | ||
NM_202853.3 | NP_974582.1 | 6 - 7 | 1116 | 77 | ||
GenBank mRNA | AF177381.1 | - | 7 - 8 | 731 | 77 | |
AY062549.1 | - | 7 - 8 | 797 | 77 | ||
BT008822.1 | - | 6 - 7 | 642 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |