Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036609.1 | NP_001031686.1 | 1 - 2 | 535 | 77 | |
NM_118240.4 | NP_193853.1 | 1 - 2 | 535 | 77 | ||
GenBank mRNA | AK316730.1 | - | 1 - 2 | 539 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |