Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118254.4 | NP_193865.2 | 1 - 2 | 364 | 136 | |
GenBank mRNA | AY065362.1 | - | 1 - 2 | 337 | 136 | |
AY122937.1 | - | 1 - 2 | 258 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |