Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118254.4 | NP_193865.2 | 4 - 5 | 745 | 129 | |
GenBank mRNA | AY065362.1 | - | 4 - 5 | 718 | 129 | |
AY122937.1 | - | 4 - 5 | 639 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |