Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341497.1 | NP_001320023.1 | 6 - 7 | 1352 | 95 | |
NM_001341498.1 | NP_001329314.1 | 6 - 7 | 1311 | 95 | ||
NM_118275.4 | NP_193886.3 | 6 - 7 | 1292 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |