Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118287.5 | NP_193898.3 | 8 - 9 | 2546 | 109 | |
GenBank mRNA | AY557186.1 | - | 8 - 9 | 2024 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |