Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036615.3 | NP_001031692.1 | 9 - 10 | 2472 | 88 | |
NM_001341510.1 | NP_001329394.1 | 9 - 10 | 3131 | 88 | ||
NM_001341511.1 | NP_001329395.1 | 10 - 11 | 2482 | 88 | ||
NM_118295.7 | NP_193906.2 | 9 - 10 | 2224 | 88 | ||
GenBank mRNA | AK229970.1 | - | 9 - 10 | 2325 | 88 | |
AY091104.1 | - | 9 - 10 | 2116 | 88 | ||
AY150491.1 | - | 8 - 9 | 1859 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |