Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341512.1 | NP_001328261.1 | 2 - 3 | 396 | 63 | |
NM_118296.4 | NP_193907.2 | 2 - 3 | 318 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |