Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341512.1 | NP_001328261.1 | 3 - 4 | 450 | 100 | |
NM_118296.4 | NP_193907.2 | 3 - 4 | 372 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |