Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203862.1 | NP_001190791.1 | 2 - 3 | 523 | 68 | |
NM_001341518.1 | NP_001320026.1 | 2 - 3 | 523 | 68 | ||
NM_118306.7 | NP_567639.1 | 2 - 3 | 670 | 68 | ||
GenBank mRNA | AF360341.1 | - | 2 - 3 | 506 | 68 | |
AY051078.1 | - | 2 - 3 | 453 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |