Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118322.4 | NP_193932.1 | 5 - 6 | 1580 | 63 | |
GenBank mRNA | AY081335.1 | - | 5 - 6 | 1442 | 63 | |
BT008899.1 | - | 5 - 6 | 1337 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |