Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341554.1 | NP_001329095.1 | 1 - 2 | 368 | 83 | |
NM_118383.4 | NP_193988.1 | 1 - 2 | 368 | 83 | ||
GenBank mRNA | BT002969.1 | - | 1 - 2 | 227 | 83 | |
BT004412.1 | - | 1 - 2 | 85 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |