Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341554.1 | NP_001329095.1 | 4 - 5 | 851 | 70 | |
NM_118383.4 | NP_193988.1 | 5 - 6 | 784 | 70 | ||
GenBank mRNA | BT002969.1 | - | 5 - 6 | 643 | 70 | |
BT004412.1 | - | 5 - 6 | 501 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |