Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118393.3 | NP_567663.1 | 1 - 2 | 361 | 69 | |
GenBank mRNA | AY059803.1 | - | 1 - 2 | 268 | 69 | |
AY087472.1 | - | 1 - 2 | 255 | 69 | ||
BT003349.1 | - | 1 - 2 | 112 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |