Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341561.1 | NP_001328982.1 | 4 - 5 | 503 | 128 | |
NM_118407.5 | NP_194009.2 | 3 - 4 | 468 | 128 | ||
GenBank mRNA | AF528063.1 | - | 3 - 4 | 341 | 128 | |
BT029176.1 | - | 3 - 4 | 221 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |