Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341568.1 | NP_001320037.1 | 4 - 5 | 669 | 92 | |
NM_001341569.1 | NP_001328192.1 | 4 - 5 | 669 | 92 | ||
NM_118424.2 | NP_194026.1 | 3 - 4 | 319 | 92 | ||
GenBank mRNA | AF312664.1 | - | 3 - 4 | 319 | 92 | |
AY142523.1 | - | 3 - 4 | 319 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |