Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203875.2 | NP_001190804.1 | 20 - 21 | 4881 | 62 | |
NM_001341570.1 | NP_001328112.1 | 20 - 21 | 4728 | 62 | ||
NM_118426.4 | NP_194028.2 | 20 - 21 | 4823 | 62 | ||
GenBank mRNA | AY823256.1 | - | 20 - 21 | 4725 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |