Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203875.2 | NP_001190804.1 | 11 - 12 | 2628 | 143 | |
NM_001341570.1 | NP_001328112.1 | 11 - 12 | 2475 | 143 | ||
NM_118426.4 | NP_194028.2 | 11 - 12 | 2570 | 143 | ||
GenBank mRNA | AY823256.1 | - | 11 - 12 | 2472 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |