Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084966.2 | NP_001078435.1 | 1 - 2 | 1095 | 93 | |
NM_001341586.1 | NP_001329248.1 | 1 - 2 | 1095 | 93 | ||
NM_001341587.1 | NP_001329247.1 | 1 - 2 | 1095 | 93 | ||
NM_118450.4 | NP_194052.2 | 1 - 2 | 1095 | 93 | ||
NM_179096.3 | NP_849427.1 | 1 - 2 | 1095 | 93 | ||
GenBank mRNA | AY142503.1 | - | 1 - 2 | 875 | 93 | |
BT001110.1 | - | 1 - 2 | 875 | 93 | ||
DQ680080.1 | - | 1 - 2 | 936 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |