Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118496.4 | NP_194096.1 | 8 - 9 | 1742 | 153 | |
GenBank mRNA | AF385710.1 | - | 8 - 9 | 1550 | 153 | |
AY081732.1 | - | 8 - 9 | 1506 | 153 | ||
AY087637.1 | - | 8 - 9 | 1546 | 153 | ||
BT000736.1 | - | 8 - 9 | 1508 | 153 | ||
U20623.1 | - | 8 - 9 | 1521 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |