Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335200.1 | NP_001323815.1 | 2 - 3 | 452 | 97 | |
NM_126419.1 | NP_178467.1 | 2 - 3 | 402 | 97 | ||
GenBank mRNA | BT010857.1 | - | 2 - 3 | 298 | 97 | |
BT020330.1 | - | 2 - 3 | 208 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |