Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148232.3 | NP_680598.2 | 5 - 6 | 2598 | 74 | |
GenBank mRNA | AK229122.1 | - | 5 - 6 | 2492 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |