Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341092.1 | NP_001319956.1 | 2 - 3 | 824 | 104 | |
NM_117725.3 | NP_193363.4 | 2 - 3 | 411 | 104 | ||
NM_148347.2 | NP_680713.2 | 3 - 4 | 620 | 104 | ||
NM_179211.3 | NP_849542.2 | 2 - 3 | 980 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |