Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341313.1 | NP_001319991.1 | 3 - 4 | 352 | 97 | |
NM_001341314.1 | NP_001319990.1 | 3 - 4 | 565 | 97 | ||
NM_001341315.1 | NP_001329781.1 | 3 - 4 | 352 | 97 | ||
NM_001341316.1 | NP_001329780.1 | 3 - 4 | 471 | 97 | ||
NM_148354.4 | NP_680720.1 | 3 - 4 | 307 | 97 | ||
NM_202841.2 | NP_974570.1 | 3 - 4 | 471 | 97 | ||
GenBank mRNA | AK317105.1 | - | 3 - 4 | 454 | 97 | |
AY065055.1 | - | 3 - 4 | 307 | 97 | ||
AY093752.1 | - | 2 - 3 | 196 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |