Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148455.2 | NP_683296.1 | 2 - 3 | 154 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |