Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333839.1 | NP_001319269.1 | 8 - 9 | 1802 | 78 | |
NM_001333840.1 | NP_001319268.1 | 8 - 9 | 1841 | 78 | ||
NM_001333841.1 | NP_001322290.1 | 8 - 9 | 1804 | 78 | ||
NM_001333842.1 | NP_001322291.1 | 7 - 8 | 1911 | 78 | ||
NM_001333843.1 | NP_001322289.1 | 8 - 9 | 1722 | 78 | ||
NM_148599.4 | NP_683440.2 | 7 - 8 | 1528 | 78 | ||
GenBank mRNA | KJ920364.1 | - | 7 - 8 | 1478 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |