Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RKF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332873.1 | NP_001322311.1 | 10 - 11 | 1069 | 101 | |
NM_102715.8 | NP_174268.7 | 10 - 11 | 1190 | 101 | ||
NM_180624.6 | NP_850955.5 | 9 - 10 | 1302 | 101 | ||
GenBank mRNA | AY091006.1 | - | 9 - 10 | 1279 | 101 | |
AY142598.1 | - | 9 - 10 | 840 | 101 | ||
FJ708642.1 | - | 9 - 10 | 840 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |