Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RKF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332873.1 | NP_001322311.1 | 5 - 6 | 710 | 97 | |
NM_102715.8 | NP_174268.7 | 5 - 6 | 831 | 97 | ||
NM_180624.6 | NP_850955.5 | 4 - 5 | 943 | 97 | ||
GenBank mRNA | AY091006.1 | - | 4 - 5 | 920 | 97 | |
AY142598.1 | - | 4 - 5 | 481 | 97 | ||
FJ708642.1 | - | 4 - 5 | 481 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |