Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102730.2 | NP_564338.1 | 1 - 2 | 1551 | 89 | |
GenBank mRNA | AF367268.1 | - | 1 - 2 | 1453 | 89 | |
AY133548.1 | - | 1 - 2 | 1343 | 89 | ||
U40341.1 | - | 1 - 2 | 1349 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |