Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102734.5 | NP_564341.2 | 14 - 15 | 1507 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |