Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198185.1 | NP_001185114.1 | 6 - 7 | 490 | 97 | |
NM_102750.5 | NP_174303.1 | 5 - 6 | 442 | 97 | ||
GenBank mRNA | AY087445.1 | - | 5 - 6 | 400 | 97 | |
BT025530.1 | - | 4 - 5 | 299 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |