Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332884.1 | NP_001320727.1 | 14 - 15 | 1944 | 63 | |
NM_102766.6 | NP_564353.1 | 14 - 15 | 1862 | 63 | ||
GenBank mRNA | AY035226.1 | - | 14 - 15 | 1392 | 63 | |
AY056419.1 | - | 14 - 15 | 1764 | 63 | ||
AY090322.1 | - | 14 - 15 | 1392 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Inventoried Probe spans exons |