Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332883.1 | NP_001320726.1 | 3 - 4 | 782 | 110 | |
NM_102766.6 | NP_564353.1 | 3 - 4 | 836 | 110 | ||
GenBank mRNA | AY035226.1 | - | 3 - 4 | 366 | 110 | |
AY056419.1 | - | 3 - 4 | 738 | 110 | ||
AY090322.1 | - | 3 - 4 | 366 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |