Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102775.4 | NP_174327.1 | 1 - 2 | 225 | 68 | |
GenBank mRNA | AY039848.1 | - | 1 - 2 | 144 | 68 | |
AY074826.1 | - | 1 - 2 | 83 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |