Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036039.2 | NP_001031116.1 | 4 - 5 | 1187 | 96 | |
NM_102777.3 | NP_174329.1 | 5 - 6 | 1125 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |